SGE™ ConnecTite™ Liners for Thermo Scientific™ GC
Designed for use with Thermo Scientific TRACE™ and FOCUS™ GCs
$110.33 - $546.20
Specifications
Diameter (Metric) Inner 0.5 cm
Diameter (Metric) Outer 0.8 cm
For Use With (Equipment) Thermo Scientific GC Trace and Focus
Length (Metric) 10.5 cm
View More Specs
Products 6
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
SG092334 View Documents Trajan Scientific America
SG092334
Connectite standard liner 5/Pk. Pack of 5 Each for $437.04
SG092335 View Documents Trajan Scientific America
SG092335
Connectite top hole liner 5/Pk. Pack of 5 Each for $546.20
SG09233401 View Documents Trajan Scientific America
SG09233401
Connectite standard liner 1/Pk. Each for $110.33
SG092336 View Documents Trajan Scientific America
SG092336
Connectite bottom hole liner 5/Pk. Pack of 5 Each for $546.20
SG09233501 View Documents Trajan Scientific America
SG09233501
Connectite top hole liner 1/Pk. Each for $137.13
SG09233601 View Documents Trajan Scientific America
SG09233601
Connectite bottom hole liner 1/Pk. Each for $137.13
Description
Description
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 8mm OD, 5mm ID; 105mm length
Specifications
Specifications
0.5 cm
0.8 cm
Thermo Scientific GC Trace and Focus
10.5 cm