Please login to your online account to display your discounted pricing

SGE™ Autosampler Syringes with 42mm Cone Tip Fixed Needle


Incorporate Diamond Syringe Technology offering significantly improved levels of durability, clarity and accuracy. SGE Autosampler Syringes with 42mm Cone Tip Fixed Needle are designed specifically for use on Agilent autosamplers.

Manufacturer: sge™  SG001822

 View more versions of this product

Catalog No. SG001822

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Needle Point Style Cone tip
For Use With (Equipment) Agilent GC Autosamplers HP7673, 7683, 7693A & 6850ALS
Model 5F-AG-0.63/0.47C 6 pack
Description Not gas tight
Connection Type Fixed needle
Diameter (Metric) Outer Needle 0.63/0.47mm
For Use With (Application) Gas chromatography
Length (Metric) Needle 42mm
Type Autosampler Syringe
Volume (Metric) 5μL
Gauge 23 to 26 ga
Length (Metric) 42mm

  • Improved solvent resistance
  • Wide temperature ranges
  • Increased operational smoothness
  • Color coded syringes by volume
  • Range of needle gauge and length options