SGE™ 15% Graphite/85% Vespel™ Straight Ferrules
Mechanically robust for long lifetimes
Manufacturer:  SGE™ SG072669
 View more versions of this product
Catalog No. SG072669
Specifications Description & Specifications
Specifications
GC
0.317cm
Agilent technologies (Hewlett-Packard) and Perkin Elmer™ detectors/injectors at atmospheric pressure
0.251 in.
Straight ferrule
15% Graphite, 85% Vespel
0.124 in.
GC
Pack of 10
0.64cm
0.74cm