missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ 15% Graphite/85% Vespel™ Straight Ferrules

Mechanically robust for long lifetimes

$68.41 - $90.07

Specifications

For Use With (Application) GC
For Use With (Equipment) Agilent technologies (Hewlett-Packard) and Perkin Elmer™ detectors/injectors at atmospheric pressure
Material 15% Graphite, 85% Vespel
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Inside Type Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inside Type Length (Metric) Width (Metric) Price Quantity  
SG072669 View Documents SGE™
SG072669
0.317cm Straight ferrule 0.74cm 0.64cm Each for $90.07
SG072657 View Documents SGE™
SG072657
0.158cm Straight ferrule 0.37cm 0.32cm Pack of 10 Each for $68.41
Description

Description

  • Form strong grip with capillary column
  • Maximum temperature: 300°C (572°F)
  • Must retighten after initial temperature cycles
  • Cannot be reused
  • Preconditioned under nitrogen for low-bleed performance

Specifications

Specifications

GC
Agilent technologies (Hewlett-Packard) and Perkin Elmer™ detectors/injectors at atmospheric pressure
15% Graphite, 85% Vespel