missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ 15% Graphite/85% Vespel™ Reducing Ferrules

Mechanically robust for long lifetimes

$65.06 - $106.00

Specifications

For Use With (Application) GC
Material 15% Graphite, 85% Vespel
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Width (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Width (Metric) Product Type Price Quantity  
SG072655 SGE™
SG072655
0.08cm 0.37cm 0.32cm Reducing ferrule Pack of 10 Each for $65.06
SG072654 SGE™
SG072654
0.05cm 0.37cm 0.32cm Reducing ferrule Pack of 10 Each for $65.06
SG072663 SGE™
SG072663
0.02cm 0.37cm 0.32cm Reducing ferrule Pack of 10 Each for $65.06
SG072659 SGE™
SG072659
0.03cm 0.37cm 0.32cm Reducing ferrule Pack of 10 Each for $106.00
SG-0726702 SGE™
0726702*
0.05cm 0.74cm 0.64cm Reducing ferrule Pack for $75.91
SG072671 SGE™
SG072671
0.08cm 0.74cm 0.64cm Reducing ferrule Pack of 10 Each for $84.20
Specifications

Description

  • Form strong grip with capillary column
  • Maximum temperature: 300°C (572°F)
  • Must retighten after initial temperature cycles
  • Cannot be reused
  • Preconditioned under nitrogen for low-bleed performance

Description & Specifications

Specifications

GC
15% Graphite, 85% Vespel