missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGC 0946, Tocris™

Highly potent and selective DOT1L inhibitor; cell permeable

$333.43 - $333.43

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C28H40BrN7O4
Formula Weight 618.57
Synonym 1-[3-[[[(2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl]methyl](isopropyl)amino]propyl]-3-[4-(2,2-dimethylethyl)phenyl]urea
Chemical Name or Material SGC 0946
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
45-411-0 R & D Systems
454110
10mg Each for $333.43
Specifications Description & Specifications

Chemical Identifiers

>99%
618.57
SGC 0946
C28H40BrN7O4
1-[3-[[[(2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl]methyl](isopropyl)amino]propyl]-3-[4-(2,2-dimethylethyl)phenyl]urea

Specifications

>99%
618.57
1-[3-[[[(2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl]methyl](isopropyl)amino]propyl]-3-[4-(2,2-dimethylethyl)phenyl]urea
C28H40BrN7O4
10mg
SGC 0946

Safety and Handling

Recommended Storage : Store at -20°C