Please login to your online account to display your discounted pricing

Sesamol, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC132691000

 View more versions of this product

Catalog No. AC132691000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 138.12
Quantity 100g
Color Off-white to beige
Melting Point 63° to 65°C
Beilstein 19, 66
Chemical Name or Material Sesamol
CAS 533-31-3
Infrared Spectrum Authentic
Synonym 3, 4-(Methylenedioxy)phenol
Packaging Glass bottle
Storage Information Darkens on storage particularly when stored in the light and at elevated temperatures
Water 0.2% max. (K.F.)
Assay Percent Range 98%
Molecular Formula C7H6O3
MDL Number MFCD00005827

Specifications

Chemical Name or Material Sesamol
CAS 533-31-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6O3
Formula Weight 138.12
MDL Number MFCD00005827
Synonym 3, 4-(Methylenedioxy)phenol