missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sesame oil, Thermo Scientific™
Manufacturer: Fisher Scientific Acros 241000010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Sesame oil | |
8008-74-0 | |
Yellow | |
255°C | |
MFCD00132243 | |
14, 8469 | |
1L | |
0.919 |
0.6mg KOH/g max. | |
0.9190g/mL | |
-5.0°C | |
Authentic | |
Glass bottle | |
1.4700 to 1.4760 | |
Solubility in water: insoluble. Other solubilities: soluble in chloroform, ether, petroleum ethers | |
Viscous Liquid |
Chemical Identifiers
8008-74-0 | |
MFCD00132243 |
Safety and Handling
EINECSNumber : 232-370-6
RTECSNumber : VU3940000
TSCA : TSCA
RUO – Research Use Only