Please login to your online account to display your discounted pricing

Sertraline hydrochloride, Tocris

5-HT re-uptake inhibitor

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material Sertraline hydrochloride
Assay Percent Range >99%
Molecular Formula C17H17Cl2N·HCl
Formula Weight 342.69
Synonym (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

239510

 
r & d systems
RDS239510
10mg Each for $201.36

239550

 
r & d systems
RDS239550
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 342.69
Quantity 10mg
Chemical Name or Material Sertraline hydrochloride
CAS 79559-97-0
Synonym (1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride
Assay Percent Range >99%
Molecular Formula C17H17Cl2N·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature