Please login to your online account to display your discounted pricing

Serotonin Creatinine Sulfate Monohydrate 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  132680010

 View more versions of this product

Catalog No. AC132680010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 405.42
Quantity 1g
Color Off-white to light beige
Merck Index 14, 8462
Infrared Spectrum Authentic
Water 3.4 to 5.4% (K.F.)
Sulfated Ash 0.05% max.
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Synonym 5-Hydroxytryptamine creatinine sulfate monohydrate
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause damage to organs.
CAS 61-47-2
MDL Number MFCD00149653
Molecular Formula H2SO4·H2O
Name Note 99%
Chemical Name or Material Serotonin creatinine sulfate monohydrate
Assay Percent Range 99%

Specifications

Chemical Name or Material Serotonin creatinine sulfate monohydrate
Name Note 99%
CAS 61-47-2
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula H2SO4·H2O
Formula Weight 405.42
MDL Number MFCD00149653
Synonym 5-Hydroxytryptamine creatinine sulfate monohydrate
Accessories

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs.

SDS & Safety Documents