Learn More
Serotonin creatinine sulfate monohydrate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 132680050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Serotonin creatinine sulfate monohydrate | |
61-47-2 | |
99% | |
H2SO4·H2O | |
5g | |
serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate | |
BKCXVJIGPVULPX-UHFFFAOYSA-N | |
3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate | |
164531 | |
99% |
99% | |
Authentic | |
Glass bottle | |
MFCD00149653 | |
14, 8462 | |
Solubility in water: freely soluble in water | |
CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O | |
405.42 | |
405.42 |
Chemical Identifiers
61-47-2 | |
405.42 | |
BKCXVJIGPVULPX-UHFFFAOYSA-N | |
164531 | |
CN1CC(=O)N=C1N.C1=CC2=C(C=C1O)C(=CN2)CCN.O.OS(=O)(=O)O |
H2SO4·H2O | |
MFCD00149653 | |
serotonin creatinine sulfate monohydrate, serotonin creatinin-sulfat-1-wasser, unii-6l522laq9u, serotonin creatine sulfate monohydrate, creatinina serotonin sulfuric acid hydrate, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-1-propanone hydrochloride, 1-propanone, 1-benzo b thien-3-yl-3-1-pyrrolidinyl-, hydrochloride, sulfuric acid compound with 2-imino-1-methylimidazolidin-4-one and 3-2-aminoethyl-1h-indol-5-ol 1:1:1 hydrate, ccris 8960, serotonincreatininesulfatemonohydrate | |
3-(2-aminoethyl)-1H-indol-5-ol;2-amino-3-methyl-4H-imidazol-5-one;sulfuric acid;hydrate |
Safety and Handling
GHS H Statement:
May cause damage to organs.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only