Fisher Science Education™ BTW1-169 Series Digital Tablet Compound Microscope
Fisherbrand™ BTW1-169 Series Digital Tablet Compound Microscope is a new line of advanced digital compound microscope combined with detachable LCD tablet technology for high school, university and laboratory use.
Manufacturer:  Fisher Science Education™ BTW1169P
 View more versions of this product
Catalog No. S23889
Description
Description
Tablet Features:
  • Pre-loaded with Motic Apps to connect, view, and share live images
  • LCD Android tablet, 20.3cm (8 in.) with 1280 X800 screen resolution
  • Built-in camera captures still and video clips
  • Mini HDMI output for high-resolution large screen viewing
  • Micro-SD card 
  • USB and wifi interface allow for direct or wireless connection
  • Built-in Motic Hub for wireless sharing
  • Coaxial focus control
Microscope Features:
  • Energy-efficient LED illumination
  • New rackless stage for ease of operation
  • Upgraded objectives for image clarity with your choice of ASC, Semi-plan, plan or plan phase options
Specifications
Specifications
Motic Imaging software, vinyl dustcover, cables, 4GB micro SD card
1.25 Abbe N.A.
Plan
LED
Microscope
BTW1-169- P
40X, 100X,400X, 1000X
Trinocular
Limited lifetime on the microscope, 1 year limited on the tablet
Documents