Honeywell™ Series 330 Face Shield Replacement Windows
Hard hat adapter for Uvex™ Bionic™ face shield is compatible with multiple hard hats and was tested with Sperian as well as major competitor styles.
Manufacturer: Honeywell™ UVS8590
Description
Uvex™ Bionic™ is a revolutionary face shield designed for rugged jobs. Extremely lightweight and highly adjustable for all-day comfort, it is perfect for workers exposed to impact, chemical splash, and airborne debris.
Specifications
Gray | |
Mounts on hard cap | |
Ratchet suspension | |
Hard Cap Adaptor |