Please login to your online account to display your discounted pricing

GE Healthcare Sera-Mag SpeedBeads™ Streptavidin magnetic beads


SpeedBeads Streptavidin exhibit a high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond much faster to a magnetic field to shorten assay times and improve precision and also move faster through viscous solutions.

Manufacturer: ge healthcare  66152104010150

 View more versions of this product

Catalog No. 09981139

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) 1μm
Storage Requirements 2° to 8°C
For Use With (Application) Immunoassays
Quantity 5mL
Concentration 1% solids (10mg/mL)

  • Available with a 3500-4500 pmol/mg biotin-binding range
  • Their "cauliflower-like" structure gives Sera-Mag beads high binding capacity
  • Combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in such immunoassay and molecular biology applications as sample preparation and assay development for genomics and proteomics