Please login to your online account to display your discounted pricing

GE Healthcare Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads

Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).

$199.32 - $5,316.00

Specifications

Concentration 1% solids (10mg/mL)
Diameter (Metric) 1μm
Storage Requirements 2° to 8°C
For Use With (Application) Affinity purification (IgA and IgG purification)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

09-981-920

 
ge healthcare
17152104011150
1mL Each for $199.32

09-981-921

 
ge healthcare
17152104010150
5mL Each for $666.00

09-981-922

 
ge healthcare
17152104010350
100mL Each for $5,316.00
Description & Specifications

Specifications

Concentration 1% solids (10mg/mL)
Diameter (Metric) 1μm
Storage Requirements 2° to 8°C
For Use With (Application) Affinity purification (IgA and IgG purification)

  • Their "cauliflower-like" structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification