Please login to your online account to display your discounted pricing

GE Healthcare Sera-Mag SpeedBeads™ Neutravidin Magnetic Beads


Capture of Biotinylated Probes. Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. Neutravidin SpeedBeads combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in immunoassay and molecular biology applications.

Manufacturer: ge healthcare  78152104011150

 View more versions of this product

Catalog No. 09981154

  • / Each
 

Description & Specifications

Specifications

Diameter (Metric) 1μm
Quantity 1mL
Storage Requirements 2° to 8°C
For Use With (Application) Immunoassays
Concentration 1% solids (10mg/mL)

  • Neutravidin beads exhibit high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond quickly to a magnetic field to shorten assay times, improve precision and move faster through viscous solutions
  • Available with a 3500-4500 pmol/mg biotin-binding range