Please login to your online account to display your discounted pricing

GE Healthcare Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophobic


Excellent for coupling oligos or proteins directly to the beads. Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.

Manufacturer: ge healthcare  65152105050450

 View more versions of this product

Catalog No. 09981125

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements 2° to 8°C
Diameter (Metric) 1μm
For Use With (Application) DNA Sample Prep
Concentration 5% solids (50mg/mL)
Quantity 1L

  • Double shell magnetite beads
  • . High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer