missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to beads. Their “cauliflower-like” structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
$674.00 - $3549.00
Specifications
Concentration | 5% solids (50mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | DNA Sample Prep, Protein Purification |
Diameter (Metric) | 1 μm |
Description
- High binding capacity
- 1.0μm diameter
- High carboxyl acid content for binding for binding DNA and proteins
- Hydrophobic bead based on crosslinking requirements for the encapsulation layer
Specifications
5% solids (50mg/mL) | |
2°C to 8°C | |
DNA Sample Prep, Protein Purification | |
1 μm |