Please login to your online account to display your discounted pricing

Alfa Aesar™ [Ser&superscript_begin;25&superscript_end;] Protein Kinase C (19-31)

For cellular and molecular biology applications

$137.00 - $212.42

Specifications

Product Type [Ser25] Protein Kinase C (19-31)
Molecular Weight 1559.84
Storage Requirements -30° to -10°C
Sequence Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66676EXD

 
alfa aesar™
J66676EXD
0.5mg Each for $137.00

AAJ66676MCR

 
alfa aesar™
J66676MCR
1mg Each for $212.42
Description & Specifications

Specifications

Product Type [Ser25] Protein Kinase C (19-31)
Molecular Weight 1559.84
Storage Requirements -30° to -10°C
Sequence Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val
Format Solid
Regulatory Status RUO
Purification Method HPLC