Please login to your online account to display your discounted pricing

Sequencing Gel Loading Dye, 3x, ACROS Organics™


Quantity 5mL

Manufacturer: fisher scientific acros  611980050

 View more versions of this product

Catalog No. AC611980050

  • / Each
 

Description & Specifications

Chemical Identifiers

Absorbance 0.40 - 0.50 AU at 252nm
DNase DNase free
Quantity 5mL
Chemical Name or Material Sequencing gel loading dye
CAS 75-12-7

Specifications

Chemical Name or Material Sequencing gel loading dye
CAS 75-12-7