Please login to your online account to display your discounted pricing

Sequencing Gel Loading Dye, 3x, ACROS Organics™

$123.25 - $123.25

Chemical Identifiers

Chemical Name or Material Sequencing gel loading dye
CAS 75-12-7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC611980050

  SDS
fisher scientific acros
611980050
5mL Each for $123.25
Description & Specifications

Specifications

Absorbance 0.40 - 0.50 AU at 252nm
DNase DNase free
Quantity 5mL
Chemical Name or Material Sequencing gel loading dye
CAS 75-12-7