Please login to your online account to display your discounted pricing
Thermo Scientific Septa for 8-425 Screw Thread Caps
Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
Manufacturer: thermo scientific  C401355
 View more versions of this product
Catalog No. 03-340-10F
  • / Pack of 100 Each
Description & Specifications
Specifications
Diameter (Metric) Outer 8mm
Type Screw Cap Septa
For Use With Hitachi™ autosamplers
Vial Closure Size 8-425mm
Material Red PTFE/Clear Silicone
Vial Closure Size 8-425mm
Thermo Scientific™ National Septa for 8-425 Screw Thread Caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of applications.
  • PTFE Disk septa are chemically inert
  • PFTE red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles