Please login to your online account to display your discounted pricing

Semi-Volatiles Mixture, 1000μg/mL in methylene chloride/benzene (3:1), Ultra Scientific


Quantity 1 x 1mL

Manufacturer: ultra scientific  801175052135

 View more versions of this product

Catalog No. USSVM82701

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1 x 1mL
Analyte or Component Names Methylene chloride (dichloromethane),benzene,acenaphthene
Solvent or Matrix Methylene chloride/benzene (3:1)
Chemical Name or Material Semi-Volatiles Mixture
CAS 000208-96-8
CAS Max % 0.08
CAS 000120-12-7
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS Max % 17.07
CAS Max % 0.08
CAS 000056-55-3
CAS 000075-09-2
CAS 000071-43-2
CAS Max % 0.08
CAS Max % 0.08
CAS 000083-32-9
CAS Max % 77.64

Specifications

Chemical Name or Material Semi-Volatiles Mixture
CAS 000075-09-2
CAS Max % 77.64
CAS 000071-43-2
CAS Max % 17.07
CAS 000083-32-9
CAS Max % 0.08
CAS 000208-96-8
CAS Max % 0.08
CAS 000120-12-7
CAS Max % 0.08
CAS 000056-55-3
CAS Max % 0.08