Please login to your online account to display your discounted pricing

Semi-Volatiles Mixture, 1000μg/mL in methylene chloride/benzene (3:1), Ultra Scientific

$562.69 - $562.69

Chemical Identifiers

Chemical Name or Material Semi-Volatiles Mixture
CAS 000075-09-2
CAS Max % 77.64
CAS 000071-43-2
CAS Max % 17.07
CAS 000083-32-9
CAS Max % 0.08
CAS 000208-96-8
CAS Max % 0.08
CAS 000120-12-7
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

USSVM82701

 
ultra scientific
801175052135
1 x 1mL Each for $562.69
Description & Specifications

Specifications

Quantity 1 x 1mL
Solvent or Matrix Methylene chloride/benzene (3:1)
Analyte or Component Names Methylene chloride (dichloromethane),benzene,acenaphthene
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 000208-96-8
Chemical Name or Material Semi-Volatiles Mixture
CAS Max % 17.07
CAS 000120-12-7
CAS Max % 0.08
CAS Max % 0.08
CAS 000056-55-3
CAS 000071-43-2
CAS 000075-09-2
CAS Max % 0.08
CAS 000083-32-9
CAS Max % 0.08
CAS Max % 77.64