Please login to your online account to display your discounted pricing

Semi-Volatile Calibration Standard, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 1mL
Packaging 2mL amber prescored glass ampule

Manufacturer: spex certiprep llc  BIGBN2

 View more versions of this product

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. BIGBN2

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1mL
Chemical Name or Material Semi-volatile Calibration Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Health Hazard 1 Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.
Packaging 2mL amber prescored glass ampule
Recommended Storage Ambient
Methods For US EPA Methods 625, 8270, CLP
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
DOT Information Nonhazardous
Solvent or Matrix MeCl:Benzene
Concentration or Composition (by Analyte or Components) Components in Methylene Chloride:Benzene at 2000μg/mL
Acenaphthene    Dimethyl Phthalate
Acenaphthylene    Di-n-butyl Phthalate
Anthracene    2,4-Dinitrotoluene
Benzo(a)anthracene    2,6-Dinitrotoluene
Benzo(a)pyrene    Di-n-octyl Phthalate
Benzo(b)fluoranthene    Fluoranthene
Benzo(g,h,i)perylene    Fluorene
Benzo(k)fluoranthene    Hexachlorobenzene
Benzyl Alcohol    Hexachlorobutadiene
bis(2-chloroethoxy)methane    Hexachlorocyclopentadiene
bis(2-chloroethyl)ether    Hexachloroethane
bis(2-chloroisopropyl)ether    Indeno(1,2,3-cd)pyrene
bis(2-ethylhexyl)phthalate    Isophorone
4-Bromophenyl phenyl ether    2-Methylnaphthalene
Butylbenzyl phthalate    Naphthalene
4-chloroaniline    2-Nitroaniline
2-chloronaphthalene    3-Nitroaniline
4-chlorophenyl-phenyl ether    4-Nitroaniline
Chrysene    Nitrobenzene
Dibenz(a,h)anthracene    N-Nitroso-di-n-propylamine
Dibenzofuran    N-Nitrosodiphenylamine
1,2-Dichlorobenzene    Phenanthrene
1,3-Dichlorobenzene    Pyrene
1,4-Dichlorobenzene    1,2,4-Trichlorobenzene
Diethyl Phthalate

Specifications

Chemical Name or Material Semi-volatile Calibration Standard
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2,4-Trichlorobenzene120-82-12000μg/mL
1,2-Dichlorobenzene95-50-12000μg/mL
1,3-Dichlorobenzene541-73-12000μg/mL
1,4-Dichlorobenzene106-46-72000μg/mL
2,4-Dinitrotoluene121-14-22000μg/mL
2,6-Dinitrotoluene606-20-22000μg/mL
2-Chloronaphthalene91-58-72000μg/mL
2-Methylnaphthalene91-57-62000μg/mL
2-Nitroaniline88-74-42000μg/mL
3-Nitroaniline99-09-22000μg/mL
4-Bromodiphenyl ether101-55-32000μg/mL
4-Chloroaniline (p-Chloroaniline)106-47-82000μg/mL
4-Chlorophenyl-phenyl ether7005-72-32000μg/mL
4-Nitroaniline100-01-62000μg/mL
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Benzyl alcohol100-51-62000μg/mL
Bis(2-chloro-1-methylethyl) ether108-60-12000μg/mL
bis(2-Chloroethoxy)methane111-91-12000μg/mL
bis(2-Chloroethyl)ether111-44-42000μg/mL
Bis(2-Ethylhexyl)phthalate117-81-72000μg/mL
Butylbenzyl phthalate85-68-72000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Dibenzofuran132-64-92000μg/mL
Diethyl phthalate84-66-22000μg/mL
Dimethyl phthalate131-11-32000μg/mL
Di-n-butyl phthalate84-74-22000μg/mL
Di-n-octyl phthalate117-84-02000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Hexachlorobenzene118-74-12000μg/mL
Hexachlorobutadiene87-68-32000μg/mL
Hexachlorocyclopentadiene77-47-42000μg/mL
Hexachloroethane67-72-12000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Isophorone78-59-12000μg/mL
Naphthalene91-20-32000μg/mL
Nitrobenzene98-95-32000μg/mL
N-Nitrosodi-n-propylamine621-64-72000μg/mL
N-Nitrosodiphenylamine86-30-62000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

Ambient