Please login to your online account to display your discounted pricing

Semaphorin 4A Mouse anti-Human, Mouse, APC, Clone: 5E3, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $433.55

Specifications

Antigen Semaphorin 4A
Clone 5.00E+03
Host Species Murine
Gene Alias Sema B, Sema4A
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123225

 
life technologies
LS17975341
25 Tests Each for $187.05

501123226

 
life technologies
LS17975342
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen Semaphorin 4A
Clone 5.00E+03
Host Species Murine
Gene Alias Sema B, Sema4A
Species Reactivity Human, Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The monoclonal antibody 5E3 reacts with human and mouse Semaphorin 4A (Sema4A). This 150kDa glycoprotein is a member of the class IV semaphorin family and exists as membrane-bound and soluble forms. Sema4A is expressed by a wide variety of tissues, including lung, kidney, spleen, testis, and mammary glands, as well as activated T cells and dendritic cells. This ligand binds to Tim-2, neuropilin-1 (Nrp1), plexin D1, and plexin B1. Sema4A plays a role in T cell activation, Th1 differentiation, and Treg function and survival.

Our testing demonstrates that the 5E3 antibody stains resting CD49b+ NK cells in mouse. However, staining of CD56+ NK cells in human is negative.