missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Selenous acid, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1085522

 View more versions of this product

Catalog No. AA1085522



Specifications

Specifications

Selenous acid
7783-00-8
H2O3Se
UN3290
ccris 5530
O[Se](=O)O
128.984
CHEBI:26642
Crystalline Aggregates
Puratronic™
Hygroscopic
100g
(metals basis)
MFCD00011331
14,8430
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenous acid
1091
128.97
99.999%
Odorless
70°C (decomposition)

Chemical Identifiers

7783-00-8
128.984
MCAHWIHFGHIESP-UHFFFAOYSA-N
1091
selenous acid
H2O3Se
MFCD00011331
ccris 5530
CHEBI:26642
O[Se](=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, CORROSIVE, INORGANIC, N.O.S.

TSCA : TSCA

Documents