missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenous acid, 97%, Alfa Aesar™

For Use As An Oxidizing Agent, As a Reagent For Alkaloids

Manufacturer:  Alfa Aesar 1257406

 View more versions of this product

Catalog No. AA1257406



Specifications

Specifications

Selenous acid
5g
97%
MFCD00011331
MCAHWIHFGHIESP-UHFFFAOYSA-N
selenous acid
128.984
CHEBI:26642
Crystalline
Colorless
7783-00-8
H2O3Se
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution
O[Se](=O)O
3.004g/cm3 at 20°C
1091
128.97
Odorless

Chemical Identifiers

7783-00-8
128.984
MCAHWIHFGHIESP-UHFFFAOYSA-N
1091
selenous acid
H2O3Se
MFCD00011331
selenious acid, selenious acid h2seo3, monohydrated selenium dioxide, unii-f6a27p4q4r, selenium dioxide, monohydrated, ccris 5530, selenious acid usp, selenious acid usp, selenious acid,selenous acid,monohydrated selenium dioxide, selenium, reference standard solution
CHEBI:26642
O[Se](=O)O
Safety and Handling

Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 6.1, Packaging Group: II

EINECSNumber : 231-974-7

Documents