Please login to your online account to display your discounted pricing

Alfa Aesar™ Selenourea, 99.97% (metals basis)

Chemical Identifiers

Chemical Name or Material Selenourea
CAS 630-10-4
Formula Weight 123.02
MDL Number MFCD00008065
UN Number UN3283
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3672003

  SDS
alfa aesar™
3672003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3672006

  SDS
alfa aesar™
3672006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 123.02
Packaging Ampule
Chemical Name or Material Selenourea
Quantity 1g
UN Number UN3283
Assay Percent Notes (metals basis)
Sensitivity Air & Moisture Sensitive
Melting Point ≈210°C (dec.)
Health Hazard 1 Danger
TSCA TSCA
Storage Note 1
  • Ampouled under argon
Color White to gray
Health Hazard 2 GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
RTECS Number YU1820000
Assay 99.97%
Health Hazard 3 GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
MDL Number MFCD00008065
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
CAS 630-10-4

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

  • Ampouled under argon