Please login to your online account to display your discounted pricing

Selenourea, 98+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213170050

 View more versions of this product

Catalog No. AC213170050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 123.02
Physical Form Crystalline Powder or Needles
Quantity 5g
Color Slightly pink to gray
Linear Formula H2NC(=Se)NH2
Chemical Name or Material Selenourea
MDL Number MFCD00008065
CAS 630-10-4
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98+%
Molecular Formula CH4N2Se
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Selenourea
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula CH4N2Se
Linear Formula H2NC(=Se)NH2
Formula Weight 123.02
MDL Number MFCD00008065
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents