missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenophene 98.0+%, TCI America™

Manufacturer:  TCI America S08291G

 View more versions of this product

Catalog No. S08291G



Specifications

Specifications

Selenophene
Yellow
1g
C4H4Se
1992
MABNMNVCOAICNO-UHFFFAOYSA-N
selenophene
136130
131.04
≥98.0% (GC)
110°C
-30°C
288-05-1
MFCD00075487
selenophen, selenophene, acmc-1cr2h
C1=C[Se]C=C1
131.047
CHEBI:30857
Clear Liquid at 20°C

Chemical Identifiers

288-05-1
131.047
MABNMNVCOAICNO-UHFFFAOYSA-N
136130
selenophene
C4H4Se
MFCD00075487
selenophen, selenophene, acmc-1cr2h
CHEBI:30857
C1=C[Se]C=C1
Safety and Handling

Safety and Handling

TSCA : No

Documents