missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Selenophene, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 271900025
Specifications
Selenophene | |
−2°C | |
Glass bottle | |
1.52 | |
Colorless to Yellow | |
2.5g | |
99% | |
MFCD00075487 | |
acmc-1cr2h, selenophen, selenophene | |
MABNMNVCOAICNO-UHFFFAOYSA-N | |
selenophene | |
136130 | |
131.04 | |
99% |
1.5200g/mL | |
Authentic | |
1.5790 to 1.5810 | |
109.9°C to 110.1°C (752.0 mmHg) | |
-38.0°C | |
288-05-1 | |
C4H4Se | |
1992 | |
Solubility in water: .. Other solubilities: azeotrope with water, soluble in all organic solvents | |
C1=C[Se]C=C1 | |
131.04 | |
CHEBI:30857 | |
Liquid |
Chemical Identifiers
288-05-1 | |
131.04 | |
MABNMNVCOAICNO-UHFFFAOYSA-N | |
136130 | |
selenophene |
C4H4Se | |
MFCD00075487 | |
acmc-1cr2h, selenophen, selenophene | |
CHEBI:30857 | |
C1=C[Se]C=C1 |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for brea
GHS Signal Word: Danger