missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium sulfide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 06810406

 View more versions of this product

Catalog No. AA6810406



Description

Description

Selenium sulfide is used as an antifungal agent to treat dandruff, seborrheic dermatitis, eborrhea and tinea versicolor. It is the active component in shampoos as an antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(decomposition)
<100°C
7488-56-4
5 g
S2Se
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
thione
24087
Selenium sulfide
Red to Brown
Powder
97%
MFCD00011227
Insoluble in water.
S=[Se]=S
143.09
143.09

Chemical Identifiers

7488-56-4
143.09
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
24087
S=[Se]=S
S2Se
MFCD00011227
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium
thione
Safety and Handling

Safety and Handling

P262-P264b-P270-P280-P301+P310-P304+P340-P311-P314-P330-P501c

H301+H331-H373

DOTInformation : DOT Class: 6.1; Packaging Group: III

EINECSNumber : 231-303-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only