Learn More
Selenium sulfide, 97%, Thermo Scientific Chemicals
$64.03 - $632.25
Chemical Identifiers
CAS | 7488-56-4 |
---|---|
Molecular Formula | S2Se |
Molecular Weight (g/mol) | 143.09 |
MDL Number | MFCD00011227 |
InChI Key | JNMWHTHYDQTDQZ-UHFFFAOYSA-N |
Synonym | selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium |
PubChem CID | 24087 |
IUPAC Name | thione |
SMILES | S=[Se]=S |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA6810406
|
Thermo Scientific Chemicals
06810406 |
5 g |
Each for $64.03
|
|
|||||
AA6810418
|
Thermo Scientific Chemicals
06810418 |
50 g |
Each for $194.40
|
|
|||||
AA6810430
|
Thermo Scientific Chemicals
06810430 |
250 g |
Each for $632.25
|
|
|||||
Description
Selenium sulfide is used as an antifungal agent to treat dandruff, seborrheic dermatitis, eborrhea and tinea versicolor. It is the active component in shampoos as an antifungal agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7488-56-4 | |
143.09 | |
JNMWHTHYDQTDQZ-UHFFFAOYSA-N | |
24087 | |
S=[Se]=S |
S2Se | |
MFCD00011227 | |
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium | |
thione |
Specifications
(decomposition) | |
Red to Brown | |
Powder | |
97% | |
MFCD00011227 | |
Insoluble in water. | |
S=[Se]=S | |
143.09 | |
143.09 |
<100°C | |
7488-56-4 | |
5 g | |
S2Se | |
selenium disulfide, selenium disulphide, selsun blue, selenium iv disulfide, selenium sulfide ses2, sebusan, selenex, selenix, selukos, lenium | |
JNMWHTHYDQTDQZ-UHFFFAOYSA-N | |
thione | |
24087 | |
Selenium sulfide |
Safety and Handling
P262-P264b-P270-P280-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : DOT Class: 6.1; Packaging Group: III
EINECSNumber : 231-303-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only