Please login to your online account to display your discounted pricing

Selenium sulfide, 94+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Selenium sulfide
Assay 52 to 55.5% (Se)
Assay Percent Range 94+%
Molecular Formula S2Se
Linear Formula SeS2
Formula Weight 143.08g/mol
MDL Number MFCD00011227
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC318141000

  SDS
thermo fisher scientific
318141000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging Glass bottle
Color Orange to red-brown
Quantity 100g
Chemical Name or Material Selenium sulfide
CAS 7488-56-4
Physical Form Orange to reddish-brown powder
Assay 52 to 55.5% (Se)
Assay Percent Range 94+%
Molecular Formula S2Se
Linear Formula SeS2
Formula Weight 143.08g/mol
MDL Number MFCD00011227
Merck Index 15, 8573
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.