Please login to your online account to display your discounted pricing

Selenium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™


Quantity 250mL
Packaging Poly Bottle

Manufacturer: spex certiprep llc  PLSE22T

 View more versions of this product

Catalog No. PLSE22T

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 250mL
Health Hazard 1 Causes skin irritation, serious eye irritation
Recommended Storage Ambient
DOT Information Nonhazardous
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Chemical Name or Material Selenium Standard
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLSE2-2Y, -2T, -2X    10,000    PLSE2-3Y, -3X
CAS Min % 94.0
CAS Max % 94.0
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS 7782-49-2
Packaging Poly Bottle

Specifications

Chemical Name or Material Selenium Standard
CAS 7782-49-2
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 94.0
Purity Grade Assurance Grade
Accessories

Causes skin irritation, serious eye irritation

Ambient