Please login to your online account to display your discounted pricing

Selenium Standard, 1000mg/L in 2% HNO3 (Assurance Grade), SPEX CertiPrep™

$81.31 - $237.26

Chemical Identifiers

Chemical Name or Material Selenium Standard
CAS 7782-49-2
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 94.0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLSE22T

 
spex certiprep llc
PLSE22T
Poly Bottle 250mL Each for $237.26

PLSE22M

 
spex certiprep llc
SPXPLSE22M
Triple-leached LDPE bottle 30mL Each for $81.31
Description & Specifications

Specifications

Quantity 250mL
Health Hazard 1 Causes skin irritation, serious eye irritation
Recommended Storage Ambient
DOT Information Nonhazardous
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Chemical Name or Material Selenium Standard
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLSE2-2Y, -2T, -2X    10,000    PLSE2-3Y, -3X
CAS Min % 94.0
CAS Max % 94.0
CAS Min % 1.0
CAS Max % 1.0
CAS 7697-37-2
CAS Min % 5.0
CAS Max % 5.0
CAS 7732-18-5
CAS 7782-49-2
Packaging Poly Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation

Ambient