missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Selenium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLSE23X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Se | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Selenium Standard | |
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00134090 MFCD00011224 | |
[Se] | |
78.97 | |
CHEBI:27568 | |
5% HNO3 |
7782-49-2,7697-37-2,7732-18-5 | |
500 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Se | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
selenium | |
6326970 | |
Assurance |
Chemical Identifiers
7782-49-2 | |
78.97 | |
BUGBHKTXTAQXES-UHFFFAOYSA-N | |
CHEBI:27568 | |
[Se] |
Se | |
MFCD00134090 MFCD00011224 | |
6326970 | |
selenium |
Safety and Handling
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only