Please login to your online account to display your discounted pricing

Selenium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$311.57 - $541.66

Chemical Identifiers

Chemical Name or Material Selenium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs
Includes:  NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PLSE2-3X

 
spex certiprep llc
PLSE23X
Triple-leached LDPE bottle 500mL Each for $541.66

PLSE2-3Y

 
spex certiprep llc
PLSE23Y
Triple-leached LDPE bottle 125mL Each for $311.57
Description & Specifications

Specifications

Quantity 500mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Solvent or Matrix 5% HNO3
Packaging Triple-leached LDPE bottle
DOT Information Hazardous
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Recommended Storage Ambient
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS 7782-49-2
CAS 7697-37-2
Chemical Name or Material Selenium Standard
CAS 7732-18-5

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Se10,000μg/mL

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient