missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium shot, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 10603A1

 View more versions of this product

Catalog No. AA10603A1



Specifications

Specifications

Selenium
99.999% (Metals basis)
MFCD00134090
alloy
[Se]
78.971
CHEBI:27568
1kg
7782-49-2
Se
Shot
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
Shot, Amorphous, 2 To 6mm (0.08 To 0.2 in.)

Chemical Identifiers

7782-49-2
78.971
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
selenium
Se
MFCD00134090
alloy
CHEBI:27568
[Se]
Safety and Handling

Safety and Handling

H301-H331-H373-H413: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. May cause long lasting harmful effects to aquatic life.

EINECSNumber : 231-957-4

Documents