Please login to your online account to display your discounted pricing

Selenium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS464500

 View more versions of this product

Catalog No. SS464-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Melting Point 0°C
Quantity 500mL
Physical Form Liquid
Color Colorless
Viscosity [NULL]
Density 1.0g/cm³
Chemical Name or Material Selenium, Reference Standard Solution
Name Note 1000ppm ±1%
Concentration 990 to 1010ppm
MDL Number MFCD00134090
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Poly Bottle
CAS 7783-00-8
CAS 7732-18-5

Specifications

Chemical Name or Material Selenium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7783-00-8
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134090
Accessories
 

SDS & Safety Documents