Please login to your online account to display your discounted pricing

Selenium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS464100

 View more versions of this product

Catalog No. SS464-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Purity Grade Certified Grade
Quantity 100mL
Physical Form Liquid
Color Colorless
Density 1.0g/cm³
Identification Pass Test
CAS 7732-18-5
Name Note 1000ppm ±1%
CAS 7783-00-8
MDL Number MFCD00134090
ChemAlert Storage Symbol Gray
Packaging Poly Bottle
Concentration 990 to 1010ppm
Chemical Name or Material Selenium, Reference Standard Solution

Specifications

Chemical Name or Material Selenium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7783-00-8
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134090
Accessories
 

SDS & Safety Documents