Please login to your online account to display your discounted pricing

Selenium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$85.47 - $135.12

Chemical Identifiers

Chemical Name or Material Selenium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7783-00-8
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134090
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS464-500

  SDS
fisher scientific company
SS464500
Poly Bottle 500mL Each for $135.12
Case of 6 Each for $810.72

SS464-100

  SDS
fisher scientific company
SS464100
Poly Bottle 100mL Each for $85.47
Case of 6 Each for $512.82
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Quantity 500mL
Physical Form Liquid
Color Colorless
Viscosity [NULL]
Density 1.0g/cm³
Chemical Name or Material Selenium, Reference Standard Solution
Name Note 1000ppm ±1%
Concentration 990 to 1010ppm
MDL Number MFCD00134090
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Poly Bottle
CAS 7783-00-8
CAS 7732-18-5
Safety and Handling