Please login to your online account to display your discounted pricing

Selenium Powder, -325 mesh, 99.5% (Metals basis), Alfa Aesar™

$41.74 - $116.32

Chemical Identifiers

Chemical Name or Material Selenium
CAS 7782-49-2
Physical Form Powder
Assay Percent Range 99.5% (Metals basis)
MDL Number MFCD00134090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1060106

  SDS
alfa aesar
1060106*
5g Each for $41.74

AA1060118

  SDS
alfa aesar
1060118
50g Each for $116.32

AA1060130

  SDS
alfa aesar
1060130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373
Health Hazard 3 GHS P Statement
P260-P261-P264-P301+P310-P304+P340-P311-P321-P405-P403+P233-P501a
Quantity 5g
Color Various
DOT Information DOT Class: 9, Packaging Group: III
Density 4.28g/cm3
Odor Odorless
Assay Percent Range 99.5% (Metals basis)
Appearance Various colors powder
Chemical Name or Material Selenium
EINECS Number 231-957-4
Physical Form Powder
MDL Number MFCD00134090
CAS 7782-49-2
Safety and Handling

Danger

GHS P Statement
P260-P261-P264-P301+P310-P304+P340-P311-P321-P405-P403+P233-P501a

GHS H Statement
H301-H331-H373