missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium Powder, -325 mesh, 99.5% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1060130

 View more versions of this product

Catalog No. AA1060130



Specifications

Specifications

Selenium
99.5% (Metals basis)
MFCD00134090
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
[Se]
78.971
CHEBI:27568
Odorless
250g
7782-49-2
Se
Various colors powder
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
Powder
Undesignated

Chemical Identifiers

7782-49-2
78.971
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
selenium
Se
MFCD00134090
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
CHEBI:27568
[Se]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373

GHS P Statement
P260-P261-P264-P301+P310-P304+P340-P311-P321-P405-P403+P233-P501a

Danger

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-957-4

Documents