missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium, 99.5+%, powder, 200 mesh, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 198070500

 View more versions of this product

Catalog No. AC198070500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

685.0°C
217.0°C
Colorless to Yellow
50 g
Se
MFCD00134090 MFCD00011224
15,8568
Solubility in water: insoluble. Other solubilities: soluble in concentrated acid, slightly soluble in diehtyl ether, almost insoluble in carbon disulfide, insoluble in ethanol
[Se]
78.97
CHEBI:27568
≥99.5%
4.81
Selenium
7782-49-2
Solution
99.5% min
Se
01,990; 04,422; 05,575; 06,507; 08,439; 10,345; 11,465; 14,281
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
BUGBHKTXTAQXES-UHFFFAOYSA-N
selenium
6326970
78.96
Glass bottle
4.8100g/mL

Chemical Identifiers

7782-49-2
78.97
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
selenium
Se
MFCD00134090 MFCD00011224
selenium, elemental, selen, hydride, atom, elemental, selenio, vandex, alloy, and compounds, gray
CHEBI:27568
[Se]
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

EINECSNumber : 231-957-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only