missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium plasma standard solution, Alfa Aesar™, Specpure™

Se 1000μg/mL

Manufacturer:  Alfa Aesar 13845AP

 View more versions of this product

Catalog No. AA13845AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Selenium
7697-37-2
HNO3
Nitric acid: 5%, Selenium: 0.15%, Water: 94.9%
[N+](=O)(O)[O-]
63.012
CHEBI:48107
23 hPa (17mm Hg) at 20°C
500mL
plasma standard solution
Clear liquid
MFCD00134090
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
944
Liquid
Undesignated

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
CHEBI:48107
[N+](=O)(O)[O-]
HNO3
MFCD00134090
944
nitric acid
Safety and Handling

Safety and Handling

H314-H333: Causes severe skin burns and eye damage. May be harmful if inhaled.

DOTInformation : DOT Class: 8, Packaging Group: III

Documents