Please login to your online account to display your discounted pricing

Selenium(IV) oxychloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Selenium(IV) oxychloride
Name Note 97%
CAS 7791-23-3
Assay Percent Range 97%
Molecular Formula Cl2OSe
Linear Formula SeOCl2
Formula Weight 165.86
MDL Number MFCD00016327
Synonym Seleninyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC318490050

  SDS
thermo fisher scientific
AC318490050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.86
Physical Form Liquid
Quantity 5g
Color Clear yellow
TSCA TSCA
RTECS Number VS7000000
EINECS Number 232-244-0
Density 2.4300g/mL
Chemical Name or Material Selenium(IV) oxychloride
CAS 7791-23-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Synonym Seleninyl chloride
Elemental Analysis 96.0% min.
Name Note 97%
Molecular Formula Cl2OSe
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00016327
Linear Formula SeOCl2
Assay Percent Range 97%

C-X bond formation reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.