Please login to your online account to display your discounted pricing

Selenium(IV) oxychloride, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC318490050

 View more versions of this product

Catalog No. AC318490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Selenium(IV) oxychloride
Name Note 97%
CAS 7791-23-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number VS7000000
EINECS Number 232-244-0
TSCA TSCA
Physical Form Liquid
Assay Percent Range 97%
Molecular Formula Cl2OSe
Linear Formula SeOCl2
Formula Weight 165.86
MDL Number MFCD00016327
Synonym Seleninyl chloride
Density 2.4300g/mL
Elemental Analysis 96.0% min.
Color Clear yellow
Quantity 5g

Specifications

Chemical Name or Material Selenium(IV) oxychloride
Name Note 97%
CAS 7791-23-3
Assay Percent Range 97%
Molecular Formula Cl2OSe
Linear Formula SeOCl2
Formula Weight 165.86
MDL Number MFCD00016327
Synonym Seleninyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents