missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Selenium(IV) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 193980100

 View more versions of this product

Catalog No. AC193980100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Selenium(IV) oxide
99.999%
White
10 g
O2Se
MFCD00003562
15, 8571
Solubility in water: 38.4g/100mL (14°C). Other solubilities: 1.11 parts/100 parts acetic acid (13.9°C),10.16 parts/100 parts methanol (11.8°C),6.67 parts/100 parts 93% ethanol (14°C),4.35 parts/100 parts acetone (15.3°C)
O=[Se]=O
110.97
110.96
Trace Metal Basis
315°C
7446-08-4
Fine Crystalline Powder
99.999%
SeO2
01,992; 02,360; 03,245; 04,422; 05,575; 06,509; 08,439; 09,409; 10,345; 11,96; 13,272; 14,177; 15,89; 17,312
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
JPJALAQPGMAKDF-UHFFFAOYSA-N
selenium dioxide
24007
100.00%
Glass bottle

Chemical Identifiers

7446-08-4
110.97
JPJALAQPGMAKDF-UHFFFAOYSA-N
24007
O=[Se]=O
O2Se
MFCD00003562
selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
selenium dioxide
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-194-7

RTECSNumber : VS8575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only